Learn More
Applied Biosystems™ MicroSEQ™ ID Microbial Identification Software, version 3.1.3
MicroSEQ ID Microbial Identification Software, version 3.1.3, allows you to easily identify and classify bacterial or fungal sequences by comparing them to a validated microbial library (available separately).
Brand:  Applied Biosystems™ A47132
Product Code. 18743634
  • 18710.00 € / Each
Explore more special offers
Description
Show More
Specifications
Specifications
MicroSEQ
Microbial Identification Software
Room Temperature
3500 Series genetic analyzers
1 each
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ MicroSEQ™ ID Microbial Identification Software, version 3.1.3 > Each