Learn More
Applied Biosystems™ MicroSEQ™ ID Purification Combo Kit v2.0, with cartridges
Shop All Applied Biosystems ProductsDescription
The MicroSEQ ID Purification Combo Kit v2.0 includes reagents for post-PCR and post-sequencing clean-up in the MicroSEQ ID microbial identification workflow. This version contains one tube of ExoSAP-IT Express Reagent for post-PCR clean-up and MicroSEQ ID Sequencing Clean-up Cartridges for post-sequencing clean-up.
ExoSAP-IT Express Reagent removes unincorporated dNTPs and primers from the post-amplification PCR mix in 5 minutes compared to 30 minutes with the original ExoSAP-IT Reagent while producing equivalent results. The MicroSEQ ID Sequencing Clean-up Cartridges provide a convenient and simple way to remove dye terminators, dNTPs, salts, and residual primers from cycle sequencing reactions. The end products can be used directly for sequencing on any Applied Biosystems genetic analyzer.
Comparability studies demonstrated equivalent results between the original ExoSAP-IT Reagent and the improved ExoSAP-IT Express Reagent coupled with the MicroSEQ ID Sequencing Clean-Up kit in the MicroSEQ ID workflow (see figure 1). Both ExoSAP-IT and ExoSAP-IT Express enzyme-treated PCR samples produced very similar sequencing results and the correct identification result.
ExoSAP-IT Express Reagent can be used downstream of the following PCR amplification kits:
- MicroSEQ 500 16S rDNA PCR Kit
- MicroSEQ Full Gene 16S rDNA PCR Kit
- MicroSEQD2 rDNA Fungal PCR Kit
MicroSEQ ID Sequencing Clean-up Cartridges can be used downstream of the following sequencing kits:
- MicroSEQ 500 16S rDNA Sequencing Kit
- MicroSEQ Full Gene 16S rDNA Sequencing Kit
- MicroSEQ D2 rDNA Fungal Sequencing Kit
Specifications
Specifications
| Content And Storage | • ExoSAP-IT Express (100 reactions), store at -5°C to -30°C • MicroSEQ ID Sequencing Clean-up Cartridges (36 cartridges), store at 2°C to 8°C |
| Format | Cartridge |
| For Use With (Application) | Sequencing |
| High-throughput Compatibility | Manual Protocols |
| No. of Reactions | 100 Reactions |
| Product Line | MicroSEQ |
| Quantity | 1 kit |
| Shipping Condition | Approved for shipment on Wet or Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.