Learn More
Applied Biosystems™ MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, with all library kits
Allows users to easily identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library. Features a validated library of over 11,000 microbial sequences (with all updated library kits), bacterial libraries of 500-bp 16S rDNA sequences, and over 9500 validated entries.
Brand:  Applied Biosystems™ A51427
Product Code. 18783624
  • 62700.00 € / Each
Explore more special offers
Description
Description
  • Allows users to easily identify and classify unidentified bacterial or fungal sequences by comparing them to a validated microbial library
  • Automatically identifies unknown specimens
  • Features a validated library of over 11,000 microbial sequences (with all updated library kits)
  • MicroSEQ™ ID sequence libraries consist of bacterial libraries of 500-bp 16S rDNA sequences and over 9500 validated entries
  • Extensive coverage of gram-negative non-fermenters, Bacillus, coryneforms, mycobacteria, and Staphylococcus
  • Libraries contain only correctly identified organisms and quality-controlled sequences derived from type strains of preeminent culture collections
  • Supports sequence polymorphisms and includes bacterial Category A biothreat agents
  • Run a plate on the SeqStudio™ Genetic Analyzer directly via the MicroSEQ™ ID Software
  • MicroSEQ™ system includes security, audit, and e-signature features
  • Qualification and validation support available
Specifications
Specifications
MicroSEQ ID Software
Microbial risk mitigation, identification
Included
1 Pk.
Includes digital download letter, license code kit, library kits (USB media and license code kits for 2018 Fungal Library, 2019 16S 500 Library, 2019 16s 500 Supplemental Library, and 16s Full Gene Library)
QST SeqStudio™ Instrument with SAE
Included
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Applied Biosystems™ MicroSEQ™ ID Software for SeqStudio™ Genetic Analyzer v1.0, with all library kits > 1 Package