missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Microtainer™ Tubes with Microgard™ Closure
Click to view available options
Closure Color:
Gold
Gray
Green
Lavender
Mint Green
Red
Includes:
Affected plain capillary tube
Affected plasma tube
Affected serum tube
Affected serum tube, UV prot.
Unaffected EDTA tube
Unaffected plasma tube
Unit Size:
Pack of 200
Description
BD Microtainer™ tubes are used for the collection, transport and processing of capillary or venous blood from skin puncture. It is ideal for use with infants, children, geriatrics and emergency patients whenever only the smallest amounts of blood are required.
- Wider diameter (8mm) for ease of collection and improved mixing
- Color-coded caps correspond to the standard Vacutainer™ color-coding system for easy identification
- Available with a variety of additives for use in various tests
- Amber-tinted tubes are available for light sensitive analytes
- Integrated collection scoop and visible fill lines
- Microgard™ “twist assist” closure reduces the risk of splatter when removing the cap to protect against exposure
- Closure can be used with a tube extender for use in 13mm x 75mm racks
Specifications
Specifications
| Color | Amber |
| Product Type | Microcollection Tube with Safety Closure |
| Additive | Lithium Heparin and Plasma Separator |
| Material | Polypropylene |
| Sterility | Non-sterile |
| Closure Type | Microgard |
| Capacity (Metric) | 400 to 600 μL |
| Diameter (Metric) Inner | 8 mm |
| Closure Included | Yes |
| Closure Color | Mint Green |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction