missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microtee PEEK 360μm with Fittings

Used to join 360μm capillary tubing together. Specifically designed with a 0.006 in. thruhole that delivers a low swept volume.

Brand:  Thermo Scientific™ P-888

Product Code. 11580924

  • 173.67 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Constructed from inert PEEK
  • Ideal for chemical resistance
  • Includes (3) F-152 and (3) P-416BLK Fittings
  • High-pressure
  • Biocompatible
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tee
Fittings
PEEK
360 &m;m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.