missing translation for 'onlineSavingsMsg'
Learn More

IDEX Health & Science, LLC MicroTight™ Unions Assembly

Used specifically to easily connect capillary tubing together

Brand:  IDEX Health & Science, LLC 02014-43

Product Code. 13040420

  • 90.85 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Our MicroTight Unions Assembly is used to easily connect capillary tubing together. True ZDV (Zero Dead Volume) allows two pieces of tubing to connect directly to each other using the included gauge and plug to ensure proper alignment. This allows for an uninterrupted fluidic path. It is for use with 1/32 in. OD Tubing and includes (2) F-126S Fittings and (1) P-553 Gauge.

  • MicroTight™ Unions Assembly
  • Used to easily connect capillary tubing together
  • True ZDV (Zero Dead Volume) allows two pieces of tubing to connect directly to each other using the included gauge and plug to ensure proper alignment
  • Allows for an uninterrupted fluidic path
  • Includes (2) F-126S Fittings and (1) P-553 Gauge
  • Thread Port Configuration Coned: 6-32
  • Material: PEEK
  • For Tubing OD: 1/32 in.
Specifications

Specifications

Union
5000 psi (345 bar)
(2) F-126 S Fittings and (1) P-553 Gauge
1/32 in. OD Tubing
1/32 in.
6-32 (Coned)
PEEK
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.