Learn More
Thermo Scientific™ Microtiter Tube Plug Strip
GREENER_CHOICE
Seal 1.2mL microtiter tubes tightly for storage using these polyethylene plug strips.
Brand:  Thermo Scientific™ 3425-11
20.20 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12328482
  • 22.45 € / Pack of 120
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Seal 1.2mL microtiter tubes tightly for storage by using Thermo Scientific™ Microtiter Tube Plug Strips.
  • LDPE non-autoclavable
  • Eight-plug strips can be easily cut apart for individual samples
  • Available sterile and nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8-cap Strip
1.2mL Microtiter Tube 8-cap plug strip
Natural
Sterile
Not Autoclavable
LDPE
10 sealed bags of twelve 8 cap strips/unit, 5 units/case
Plug Strips