missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microtiter Tube Plug Strip
GREENER_CHOICE

Seal 1.2mL microtiter tubes tightly for storage using these polyethylene plug strips.

19.65 € - 22.45 €

Specifications

Autoclavable Not Autoclavable
Description 1.2mL Microtiter Tube 8-cap plug strip
Color Natural
For Use With (Equipment) Plug Strips
View More Specs

Products 2
Product Code Brand Description Sterility Color Packaging Price Quantity & Availability  
Product Code Brand Description Sterility Color Packaging Price Quantity & Availability  
12328482
GREENER_CHOICE
View Documents
Thermo Scientific™
3425-11
1.2mL Microtiter Tube 8-cap plug strip Sterile Natural 10 sealed bags of twelve 8 cap strips/unit, 5 units/case
22.45 €
Pack of 120
Estimated Shipment: 03-03-2025
to see stock.
Add to basket
 
12368202
GREENER_CHOICE
View Documents
Thermo Scientific™
3426-11
1.2mL Microtiter Tube 8-cap plug strip Non-sterile Natural 10 sealed bags of twelve 8 cap strips/unit, 5 units/case
19.65 €
Pack of 120
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Seal 1.2mL microtiter tubes tightly for storage by using Thermo Scientific™ Microtiter Tube Plug Strips.

  • LDPE non-autoclavable
  • Eight-plug strips can be easily cut apart for individual samples
  • Available sterile and nonsterile

Specifications

Specifications

Not Autoclavable
1.2mL Microtiter Tube 8-cap plug strip
Natural
Plug Strips
Videos
SDS
Documents

Documents

Product Certifications