Learn More
Thermo Scientific™ Microtiter Tubes and Racks
GREENER_CHOICE
Product Code. 11937675 Shop All MBP Molecular BioProducts Products
Click to view available options
Product Type:
Frosted MicroTiter Tube
Microtiter Tube
Sterility:
Non-sterile
Sterile
Format:
Bulk
Robotic Rack
Unit Size:
Pack of 1000
Pack of 10000
Pack of 960
Product Code. 11937675
Brand: Thermo Scientific™ 3496
Seal microtiter tubes tightly for storage with these polyethylene plug strips -- for use with 1.2mL MCT titertube racks.
Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.
Microtiter Tubes and Caps
  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight-plug strips can be easily cut apart for individual samples
  • 1.2mL microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
  • Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
  • Microtiter tube system is available in bulk packaging or with a SBS standard footprint
  • Robotic compatible rack can easily integrated into automated liquid handling workstations
Highest Quality
  • Rigorous testing to ensure sample integrity and security
Packaging Options
  • Microtiter tubes and caps are available sterile and nonsterile
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Certified free of RNase, DNase, DNA and pyrogens
Color Natural
Volume (Metric) 1.2 mL
Quantity 96 tubes/tray 10 trays/pack, nonsterile
Closure Material Polyethylene (Plug)
Product Line Thermo Scientific
Sterility Non-sterile
Format Robotic Rack
Product Type Microtiter Tube