missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Microtiter Tubes and Racks
GREENER_CHOICE

Seal microtiter tubes tightly for storage with these polyethylene plug strips -- for use with 1.2mL MCT titertube racks.

37.57 € - 352.74 €

Specifications

Color Natural
Volume (Metric) 1.2 mL
Material Polyethylene (Plug)
Product Type Microtiter
View More Specs

Products 5
Product Code Brand Description Color Price Quantity & Availability  
Product Code Brand Description Color Price Quantity & Availability  
12358202
View Documents
Thermo Scientific™
3424-11
1.2mL Frosted MicroTiter Tube Robotic Rack, Nonsterile Natural This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11579944
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
3487
1.2mL Microtiter Tube Robotic Rack, Sterile Natural
49.75 €
Pack of 960
Please to purchase this item. Need a web account? Register with us today!
11937675
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
3496
1.2mL MicroTiter Tube Robotic Rack, Nonsterile Natural
42.62 €
Pack of 960
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
 
10707994
View Documents
Thermo Scientific™
3498
1.2mL Frosted MicroTiter Tube Bulk, Nonsterile Natural This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11539954
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
3492
1.2mL Microtiter Tube, Bulk, Nonsterile Natural
37.57 €
Pack of 1000
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Seal microtiter tubes tightly for storage with polyethylene Thermo Scientific™ Microtiter Tube Plug Strips — for use with 1.2mL MCT Titertube Racks. The 1.2mL microtiter tubes offer additional volume capacity for sample dilution. These are available in a pre-racked 8 x 12 format.

Microtiter Tubes and Caps

  • Polyethylene plug strip seals the microtiter tubes tightly for storage
  • Eight-plug strips can be easily cut apart for individual samples
  • 1.2mL microtiter tubes have additional volume capacity for sample dilution and are ideal for HTLV-III testing, RIA, EIA, PCR, pharmaceutical quality control and blood bank sample freezer storage
  • Frosted writing areas on each tube and an alphanumeric rack simplify organization and identification of specific samples
  • Microtiter tube system is available in bulk packaging or with a SBS standard footprint
  • Robotic compatible rack can easily integrated into automated liquid handling workstations

Highest Quality

  • Rigorous testing to ensure sample integrity and security

Packaging Options

  • Microtiter tubes and caps are available sterile and nonsterile
Specifications

Specifications

Natural
1.2 mL
Polyethylene (Plug)
Microtiter
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.