missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labcon™ Microtube Floating Storage Racks
Available in 24- or 48-place arrays with or without lids.
16.95 € - 26.45 €
Product Code | Brand | Size | Closure Included | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Closure Included | Price | Quantity & Availability | ||||
17537896
|
Labcon™
3035-500-000-9 |
24-place | None |
26.45 €
Pack of 10 |
Estimated Shipment: 28-08-2024
Log in to see stock. |
||||
17547896
|
Labcon™
3036-220-000-9 |
24-place | Clear Lid |
16.95 €
Pack of 10 |
Estimated Shipment: 13-09-2024
Log in to see stock. |
||||
17527896
|
Labcon™
3035-230-000-9 |
48-place | Clear Lid |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Holds 1.5, 1.7, or 2 mL microtubes
- Freezable to -80°C
- Made in USA
Specifications
Autoclavable | |
White | |
Polypropylene |