missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Microvascular Growth Supplement (MVGS)
Microvascular Growth Supplement (MVGS)
Brand: Gibco™ S00525
This item is not returnable.
View return policy
Description
A sterile, concentrated (20X) solution intended for use with Medium 131 to culture human microvascular endothelial cells. Each 25 ml bottle of MVGS contains all of the growth factors, hormones, and tissue extracts necessary for the culture of human microvascular endothelial cells, and is the correct amount of supplement for a 500 ml bottle of Medium 131. When a 500 ml bottle of medium is supplemented with MVGS, the final concentrations of the components in the supplemented medium are:Specifications
minus 20 degrees C | |
Growth Supplement | |
Cascade Biologics™ | |
Dry Ice |
20 X | |
Sterile | |
2 Preps | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction