Learn More
Honeywell Safety Products™ Millenia Safety Glasses Product Code.: 12682623
In Stock
to see stock.
Add to basket
Honeywell Safety Products™ Millenia Safety Glasses
Product Code. 12682623
Blue, Clear, Flexicord retainer, Impact-resistant, Anti-fog, UV-treated
Click to view available options
Frame Color:
Black
Blue
Silver
Lens Tint:
Clear
Silver Mirror
Yellow
Includes:
Adjustable, breakaway Flexicord retainer
Flexicord retainer
Lens Coating:
Impact-resistant, Anti-fog, UV-treated
Impact-resistant, Scratch-resistant, UV-treated
Product Code. 12682623
Brand: Honeywell Safety Products™ 1000006
In Stock
to see stock.
Edge
Add to basket
Add to basket
Virtually weightless eye protection for the active work environment. Honeywell Safety Products™ Millenia Safety Glasses block UV and provide protection for mechanical work including grinding, turning and milling.
Plano lens fashion safety spectacle, wrap around design with abrasion resistant lens and integral side shields. Available in a range of frame and lens colors.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number EC1131, EC Attestation Number EC1131
Lens Coating Impact-resistant, Anti-fog, UV-treated
Lens Tint Clear
Product Type Safety Glasses
For Use With (Application) Administration, Automotive, Energy or Electricity, Laboratory, Medical and Pharmaceutical and Wood industries
Frame Color Blue
Includes Flexicord retainer
No. per Pack 1 Ea.
Weight (Metric) 32 g
Lens Material Polycarbonate
Frame Material Nylon
UV Protection UV400
Product Title
Honeywell Safety Products™ Millenia Safety Glasses >