missing translation for 'onlineSavingsMsg'
Learn More

Honeywell™ Miller H-Design™ Hi-Vis Vest Safety Harness

Product Code. 15646998
missing translation for 'orderingAttributeHoverText'
:
Size 3, yellow
This item is not returnable. View return policy

Product Code. 15646998

missing translation for 'mfr': Honeywell™ 1032919

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Comfortable, high-visibility vest and 2-point safety harness.

  • 2-point Miller H-Design™ harness with stretch webbing, 2 loops, auto buckles.
  • High-visibility yellow textile vest with reflective tape certified to EN ISO 20471 (2013)
  • Side zippers with double cursers allows a perfect fit to any body shape and adjustment to summer and winter clothing
  • Inner zippers for easy access to the harness for making adjustments
  • Removable vest easy to fix to the harness thanks to its quick clip system (push buttons)
  • Vest suitable to any Miller H-Design harness:
    • 2 large front slits allow the height adjustment of the sternal anchor point and the chest straps.
  • 1 slit at the back with Velcro fastener to sustain the rear D-ring.
TRUSTED_SUSTAINABILITY

Specifications

Buckle Style Auto buckles
Certifications/Compliance EN 361, EN ISO 20471 class 2 (2013), EN ISO 13688 (2013), OEKO-TEX standard 100
For Use With (Application) Construction, fall prevention
Size 3
Style H-design
Color Yellow
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.