missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ LB Broth, High Salt, Granulated
For propagation and maintenance of E. coli.
83.25 € - 258.00 €
Specifications
Media Type | Miller Luria-Bertani Broth |
---|---|
Product Type | Microbiology Media |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12801660
|
Fisher BioReagents™
12801660 |
500 g |
83.25 €
500g |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
12891650
|
Fisher BioReagents™
12891650 |
2000 g |
258.00 €
2kg |
Estimated Shipment: 10-12-2024
Log in to see stock. |
|||||
Description
- Material specially granulated to reduce dust and increase solubility rate
- Sterilize by autoclaving
Specifications
Miller Luria-Bertani Broth | |
Microbiology Media |