missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Merck Millex™-GP Syringe Filter, PES, 0.45 μm
Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses
Brand: Merck SLHP033NK
2190.00 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Ideal for preparing 10–100mL samples prior to chromatography or other instrumental analyses
Overview of Non-Sterile Millex™ Syringe Filters with PES Membrane:- Non-sterile Millex™ syringe filters with Millipore Express™ PLUS polyethersulfone (PES) membrane provide fast filtration of aqueous and mild organic solutions.
- Available in 0.22 μm and 0.45 μm pore sizes and two diameters to suit your application needs
- Low protein binding to minimize interaction with your sample and maximize recovery
- Polypropylene housing, offering a low level of extractables and broad chemical compatibility
Applications: Buffer Filtration, Clarification of Aqueous Solutions
Spécification
Single-use | |
PES | |
Non-sterile | |
Analytical sample prep | |
Male Luer slip | |
1000Pack | |
<200 mL | |
33 mm |
Millipore Express PLUS (PES) | |
0.45 μm | |
4.5 cm2 | |
PP | |
Millex™ | |
Syringe Filter | |
33 mm |