Learn More
Merck Millicell Hanging Cell Culture Insert
Allows attachment or suspension cells to access media from both their apical and basolateral sides.
Brand:  Merck MCRP24H48
 View more versions of this product
Product Code. 15449779
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Cell growth, structure, and function more closely mimic what occurs in vivo
  • In addition, Millicell inserts make it possible to study both sides of the cell monolayer
  • The inserts are easily prepared for SEM and TEM visualizing techniques, and they are compatible with cellular and/or fluorescent stains
  • For co-culturing and permeability assays
  • Unique design allows easier basolateral access than other hanging inserts with less risk of contamination
  • Available in 5 pore sizes and 3 diameters, including a 1 μm pore size that is optically transparent for better visualization by microscopy
  • PET Membrane (polyethylene terephthalate) for growth of attachment-dependent cells without matrix
  • Individually blister packaged
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PET (Polyethylene Terephthalate) Filter Code
1 μm
50°C Max. Operating Temperature
6.5 mm
24-well Plates
Sterile
Polystyrene
Cell Culture Insert
Cell Attachment, Cell Culture, Cell Differentiation, Immunocytochemistry
48/Pk
Clear
24