Learn More
Merck Millicell HY 3-layer Cell Culture Flask
Sterile, high-yield, multi-layerd cell culture flask. Millicell HY 3-layer Cell Culture Flask provides a consistent, quality growth environment across all layers, with the same volume of media in each layer.
Brand:  Merck PFHYS0616
Product Code. 11795383
  • 904.00 € / Pack of 16
Estimated Shipment: 28-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Consistent, high cell yields and viability
  • Same media requirements (volume per unit area) as traditional T-flasks
  • Uniform culture conditions without media leaking or spillover between layers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polystyrene
222 mm
16
45°C Max.
Sterile
32 mm
3 layers
2/Bag
12 cm