Learn More
Merck Millicell™ Receiver Trays
Comes with lid
Brand:  Merck PSSW010R5
 View more versions of this product
Product Code. 10347753
  • 180.00 € / Pack of 5
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Designed to support cell attachment, growth, and differentiation for a variety of applications
  • 24-well feeder tray incorporates a tear-drop well design to reduce the incidence of air bubbles that can form under the membrane during plate assembly, ensuring that cell monolayers are getting the proper nutrients
  • Single-well feeder tray utilizes baffles to reduce media leakage and contamination
TRUSTED_SUSTAINABILITY
Specifications
Specifications
With Lid
Polystyrene
Sterile
1
5 μm
Clear
Rectangular
Receiver Tray
Lid, Single Well Receiver Tray