missing translation for 'onlineSavingsMsg'
Learn More

Millon's reagent, for the detection of Tyrosine, Thermo Scientific Chemicals

Product Code. 15550574
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10045-94-0
Molecular Formula HgN2O6
Molecular Weight (g/mol) 324.60
MDL Number MFCD00011038
InChI Key ORMNPSYMZOGSSV-UHFFFAOYSA-N
Synonym mercuric nitrate, mercury nitrate, mercury ii nitrate, citrine ointment, mercury dinitrate, millon's reagent, mercury pernitrate, mercury 2+ nitrate, nitrate mercurique french, unii-2fmv9338bw
PubChem CID 24864
IUPAC Name mercury(2+);dinitrate
SMILES [Hg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Quantity 100 mL
Linear Formula Hg(NO3)2
Formula Weight 324.6
Specific Gravity 1.4
Density 1.4000g/mL
Chemical Name or Material Millon's reagent
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Millon's reagent
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H310-Fatal in contact with skin.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H290-May be corrosive to metals.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'