missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Miltex™ Adson Dressing Forceps
Manufactured from stainless steel; hardened, tempered, and polished for a high-priced finish without the added cost
Brand: Cole-Parmer™ 10918-22
This item is not returnable.
View return policy
Beschreibung
- Dissection-grade instruments are manufactured from stainless steel; hardened, tempered and polished for a high-priced finish without the added cost
- Well-suited for less critical applications with tight budgets such as necropsies
Spezifikation
Curved | |
Stainless Steel | |
Adson Dissecting-grade Dressing Forceps | |
Forceps |
4.75 in. | |
12.065 cm | |
Dissecting |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Cole-Parmer™ Miltex™ Adson Dressing Forceps > Miltex™ Adson Dressing Forceps
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur