Learn More
Thermo Scientific™ Minerals Modified Glutamate
Enrich and identify members of the coliform group of bacteria in food and water samples with these ready-prepared bottles.
46.45 € - 52.05 €
Specifications
Color | Purple, Clear |
---|---|
Content And Storage | 2 to 25°C |
Format | Universal Bottle |
Manufacturing Origin | United Kingdom |
Product Type | Agar |
Product Code | Brand | Quantity | Media Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Media Type | Price | Quantity & Availability | ||||
12726989
|
Thermo Scientific™
BO0541E |
24/Pk. | MMGA (Bottled) |
46.45 €
Pack of 24 |
Estimated Shipment: 24-12-2024
Log in to see stock. |
||||
12736989
|
Thermo Scientific™
BO0542E |
24/Pk. | MMGA 2X Strength + DurhamTube |
52.05 €
Pack of 24 |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
Description
Enrich and identify members of the coliform group of bacteria in food and water samples with Thermo Scientific™ Oxoid™ Minerals Modified Glutamate ready-prepared bottled medium.The mineral content of the medium allows for the superior cultivation and enumeration of coliforms.
Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of Minerals Modified Glutamate Medium.
- Convenient: pre-filled bottles are ready for immediate use.
- Simple: easy-to-use with minimum training.
- Long shelf life: up to 12 months.
- Superior performance over MacConkey Broth in the detection of E. coli2.
- The medium of choice for detection of small numbers of E. coli in chlorinated water.
- Effectively isolates E. coli from sea water.
Minerals Modified Glutamate Medium was developed by Oxoid Laboratories and Metropolitan Water Board Laboratories, and is based upon the formulation described by Gray1
Specifications
Purple, Clear | |
Universal Bottle | |
Agar | |
24/Pk. |
2 to 25°C | |
United Kingdom | |
24 universal bottles with screw cap closures in a box |
Safety and Handling
missing translation for 'shelfLife' : 52 Weeks