Learn More
Thermo Scientific™ Mini Capillary Unions
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Brand:  Thermo Scientific™ 290GU498
 View more versions of this product
Product Code. 10311155
  • 68.37 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Mini Capillary Unions are for use with capillary GC columns.
Kit contains one union and five ferrules
Includes:
Union, 1; Ferrules, 5
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Capillary Union
For 0.32 mm I.D. Capillary
0.22 to 0.32 mm ID Capillary GC Column