Learn More
Ohaus™ Mini Digital Vortex Mixer VXMNDG
OHAUS™ Mini Digital Vortex Mixer VXMNDG is built sturdy to provide a stable and reliable vortexing action.
Brand:  Ohaus™ 30392124
413.37 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Product Code. 15702239
  • 528.00 € / Each
Estimated Shipment: 15-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Sturdy construction for stable and reliable vortexing action
  • Ideal for applications that demand repeatable results
  • Microprocessor control maintains set speed for strong, consistent mixing action
  • Timer will display elapsed time or, when programmed to user defined time limit, the unit will shut off when time reaches zero
  • Two modes of operation; continuous mode when using accessory attachments or touch mode which activates mixing when depressing the cup head
  • LED displays for speed and time
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4.9 mm
320 V, 0.6 A, 50/60 Hz
21.00 x 12.30 cm
16.5 cm
21.00 cm
12.30 cm
500 to 2500 rpm
0.6 A
6.50 in.
8.30 in.
4.80 in.
VXMNDG