Learn More
Neogen Mini Flip-Top Vial
Click to view available options
Includes:
9 mL Letheen Broth
9 mL Maximum Recovery Diluent
Buffered Peptone Water Broth
Unit Size:
Pack of 100
Beskrivelse
- Hinged lid can be opened with one hand and stays open for easy sample additions.
- Decontaminated through gamma irradiation and ready for use with no additional blending required.
- Flat bottom tube stands on its own, no holder required.
- Leak resistant vial helps prevent leakage during shipping.
- 2 year shelf life from date of manufacture provides long-term readiness.
Tekniske data
Tekniske data
| Includes | Buffered Peptone Water Broth |
| Quantity | 100/Cs. |
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion