Learn More
Buerkle™ Adapter for Buerkle™ MiniSampler
Manufactured from PTFE. Buerkle™ MiniSampler Adapter fits glass thread GL45. For use with Buerkle™ MiniSampler PTFE.
Brand: Buerkle™ 5305-0209
188.20 EUR valid until 2024-12-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Manufactured from PTFE
- Fits glass thread GL45
Specifications
Adapter | |
PTFE | |
Fits glass thread GL 45 |