missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Mini Vacuum Desiccator

The small size of this desiccator allows quick vacuum creation for rapid, repeated degassing of multiple samples.

Brand:  Bel-Art™ SP Scienceware™ F41099-0000

Product Code. 12643995

  • 117.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Vacuum rating of 740mm Hg (29 in.)
  • Clear polycarbonate top for easy viewing of contents; white polypropylene bottom
  • Stopcock with 2mm Teflon™ PTFE plug accepts tubing with ¼ in. to ⅜ in. I.D.
  • Peripheral flange around the bottom fits into two snap locks and seals the two halves against a Buna-N O-ring
Specifications

Specifications

Translucent
Stopcock with 2mm Teflon PTFE plug
0.56 L
5 in.
17.5 cm
Polycarbonate Top, Polypropylene Bottom
0.02 cu. ft.
12.6 cm
6.75 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.