missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Mini Vortexer

Fisherbrand™ Mini Vortexer is designed with a unique cup head that prevents liquids from entering the housing, prolonging motor life.

Brand:  Fisherbrand™ BV202-GRN-E-FIS

 View more versions of this product

Product Code. 16343196

  • 203.00 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Compact
  • Powerful mixing
  • Touch activation
Specifications

Specifications

4.5 mm
100/240 V, 50/60 Hz, 1 A, ≤50 W
EU & UK
3.8 x 3.8 in.
6.35 cm
9.65 cm
9.65 cm
0.5 kg
Cup Head
Touch
2800 rpm
50 mL
ABS Plastic Base, TPR Rubber Platform
7 in.
3.8 in.
3.8 in.
1 lb.
Green
DC
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.