Learn More
Schmidlin™ Mini Whisper Nitrogen Generator
This nitrogen generator produces a continuous supply of high purity nitrogen.
Brand:  Schmidlin™ DB-MWN2-10
Encompass_Preferred
Product Code. 17768889
  • 8952.90 € / Each
Explore more special offers
Description
Description
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Flow rate at 8 barg (116 psig)
  • Purity: LC/MS grade
  • Proprietary membrane technology
  • No noise, no moving parts and no electricity
  • Ideal for all LC/MS applications
Specifications
Specifications
18 cm
73.5 cm
7 bar
34.8 cm
>99%
12 mL/min.
35°C
13.7 in.
Inlet: 3/8 in., Outlet: 1/4 in.