Learn More
Huber™ Minichiller
Minichillers are a space-saving, low-cost and environmentally friendly cooling solution for numerous laboratory applications. By saving cooling water, Minichillers pay for themselves after only a short time.
Brand:  Huber™ 3006.0092.98
 View more versions of this product
Product Code. 15733047
  • 7440.00 € / Each
Estimated Shipment: 16-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The compact Minichillers are an economic and environmentally friendly cooling solution for many laboratory applications. A calculation of water and waste water costs shows that a Minichiller can save up to 48,000 L of water in a working week and due to its low procurement price, return on investment is achieved within a very short time.
  • The units have small footprints, only requiring 225 x 360 mm on the laboratory bench and are therefore suitable for cooling reactor blocks, vacuum pumps, rotary evaporators or heat exchangers.
  • Minichillers allow stable application conditions due to constant values for temperature, pressure and throughflow quantities.
  • They are also perfect for continuous operation at ambient temperatures up to + 40°C.
  • Despite the economic purchase price the coolers are well equipped and have a large temperature indicator, fill level indicator, as well as status LEDs for the pump, cooling and heating.
  • A 1 kW heater is optionally available with which the temperature range can be extended to +100°C.
  • Minichillers are available as both air and water cooled models and use natural refrigerants.
Specifications
Specifications
USB (Device), RS232
Pressure-suction Circulation Pump
Recirculating Chiller
Natural Refrigerant
220/240 V
0.3 kW at 15°C, 0.2 kW at 0°C, 0.14 kW at -10°C, 0.07 kW at -20°C
-20°C to 100°C
Minichiller 300w-H OLE
200 to 240 V, 50/60 Hz
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products