Learn More
Greiner Bio-One MiniCollect™ Tube, Red Cap, CAT Serum Clot Activator, Non-Sterile
Includes inner wall coated with microscopic silica particles to activate coagulation.
Brand:  Greiner Bio-One 450534
Product Code. 15788169
  • 1515.00 € / Pack of 500
Explore more special offers
Description
Description
  • The inner wall of the serum tube is specially coated with microscopic silica particles, which activate the coagulation process.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red Cap
Non-sterile
Yes
500/Pk.
CAT Serum Sep Clot Activator
0.5 mL, 1.0 mL
Red