missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alpha Packaging™ Minigrip™ Polyethylene Resealable Bags
Manufactured from polyethylene. Alpha Packaging™ Minigrip™ Polyethylene Resealable Bags are ideal for sending samples.
Brand: Alpha Packaging™ 12390549
Description
- Resealable section for the specimen
- Plain bag; no biohazard warning label
- Opening along width
- 50μm thickness.
- Polyethylene
- Three write-on bars for labelling
Specifications
Resealable Bag | |
Ideal for storing stationery and sending samples | |
50 μm | |
380 mm |
Clear | |
Polyethylene | |
100 Pack | |
510 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Alpha Packaging™ Minigrip™ Polyethylene Resealable Bags > 380W x 510mmD
Spot an opportunity for improvement?Share a Content Correction