Learn More
Lovibond™ Water Testing Minikit
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
Brand:  Lovibond™ 414140
 View more versions of this product
Product Code. 10228330
  • 59.90 € / Each
Explore more special offers
Description
Description
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
  • Easy operation and exact reagent dosing
  • High accuracy
  • Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
  • Unrestricted shipment
  • Safe storage
Specifications
Specifications
Alkalinity-P, CaC03
20 to 500mg/L
Tablet count
Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions.
Approximately 40 Tests
Minikit water testing kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lovibond™ Water Testing Minikit > Analytes: Alkalinity-P