Learn More
Cytiva Minimate™ Tangential Flow Filtration (TFF) Capsule with Omega™ Membrane
Concentration and diafiltration (desalting or buffer exchange) processes can be performed on the same system with minimal user intervention.
Brand:  Cytiva OA500C12
 View more versions of this product
Product Code. 17154771
  • 444.00 € / Each
Explore more special offers
Description
Description
  • Efficient processing
  • This product is not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PES
Non-sterile
1/Pk.
500 kDa
50 cm2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Minimate™ Tangential Flow Filtration (TFF) Capsule with Omega™ Membrane > 500 kDa