missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Minimum Essential Medium Eagle, Liquid

Incorporates higher amino acid concentrations for the cultivation of fastidious cells

37.80 € - 66.25 €

Specifications

Sterility Sterile
Quantity 500 mL
View More Specs

Products 3
Product Code Brand Product Type With Additives Without Additives Quantity Price Quantity & Availability  
Product Code Brand Product Type With Additives Without Additives Quantity Price Quantity & Availability  
11440335
View Documents Promotion Details
MP Biomedicals™
091210254
Minimum Essential Medium Eagle's Earle's Balanced Salt Solution (EBSS), Sodium Bicarbonate No Glutamine 500 mL
37.80 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11450415
View Documents Promotion Details
MP Biomedicals™
091641454
Minimum Essential Medium Eagle's (modified) Sodium Bicarbonate, Earle's Balanced Salt Solution (EBSS) No Glutamine, No Cysteine, No Cystine, No Methionine 500 mL
55.90 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Currently Unavailable: We have no information when this product will be available
View alternative products
11490335
View Documents Promotion Details
MP Biomedicals™
091213254
Minimum Essential Medium Eagle's Hanks’ Balanced Salt solution (HBSS), Sodium Bicarbonate No Glutamine 500 mL
66.25 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MEM, originally prepared by Harry Eagle, is one of the most popular cell culture media. Upon his attempts to cultivate normal mammalian fibroblasts and certain HeLa cell subtypes, it was revealed that the nutritional needs of these cell types could not be met by BME. Further studies led to the development of MEM from MP Biomedicals incorporating specific modifications such as higher amino acid concentrations for the cultivation of fastidious cells.

  • MEM from MP Biomedicals may be used to support the growth of cells in monolayers, in suspension and wide variety of other cell types with proper supplementation.
  • Available with either Earle's or Hank's salts.
Specifications

Specifications

Sterile
500 mL
Videos
SDS
Documents

Documents

Product Certifications

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.