Learn More
Gilson™ Minipuls Evolution™ Laboratory Peristaltic Pump
Speed control module for liquid transfer applications.
Brand:  Gilson™ F110701
Product Code. 12386112
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Safe and durable design
  • Stainless-steel rollers for great durability and chemical resistance
  • Low operating noise
Specifications
Specifications
Liquid transfer applications
Speed control module
169 mm
Line Frequency: 50 to 60 Hz / Line Voltage: 100 to 240 VAC / Power consumption:full load 48 w at 220 V – no load 35 w at 220 V
50 to 60 Hz line frequency
Drive module, Standard accessory package
164 mm
2.4 kg
100/240 VAC