missing translation for 'onlineSavingsMsg'
Learn More

Sartorius Stedim Biotech™ Minisart™ PES Aqueous Filtration Filter

Sartorius Stedim Biotech™ Minisart™ PES Aqueous Filtration Filter is a premium quality, PES filter designed for optimal flow rate liquid filtration.

Brand:  Sartorius Stedim Biotech™ 1776D----------Q

Additional Details : Weight : 0.60000kg

Product Code. 15698058

  • 855.00 € / Pack of 500
In Stock
to see stock.
Add to basket

Description

Description

  • Premium filters for fail-safe filtration
  • Can be used for sterile filtration of liquids
  • Optimized for high flow rates
  • Can be sterilized by autoclaving or EO
Specifications

Specifications

White
PES
Non-sterile
Polypropylene
15 mm
Male luer slip connection
0.22 μm
Liquid filtration
500/pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers