Learn More
Minocycline hydrochloride, MedChemExpress
MedChemExpress Minocycline hydrochloride is an orally active, potent and BBB-penetrated semi-synthetic tetracycline antibiotic.
Brand:  Medchem Express HY-17412
Encompass_Preferred
 View more versions of this product
Product Code. 16427296
  • 105.30 € / 100mg
Explore more special offers
Description
Description
Minocycline hydrochloride is a hypoxia-inducible factor (HIF)-1α inhibitor. Minocycline hydrochloride shows anti-cancer, anti-inflammatory, and glutamate antagonist effects. Minocycline hydrochloride reduces glutamate neurotransmission and shows neuroprotective properties and antidepressant effects. Minocycline hydrochloride inhibits bacterial protein synthesis through binding with the 30S subunit of the bacterial ribosome, resulting in a bacteriostatic effect.
Specifications
Specifications
Minocycline hydrochloride
Yellow
C23H28ClN3O7
H2O : 10 mg/mL (20.25 mM; Need ultrasonic) ∣DMSO : 0.49 mg/mL (0.99 mM; Need ultrasonic and warming)
493.94
98.47%
Powder
13614-98-7
Research
100 mg
OC1=C(C(N)=O)C([C@@]2(O)C(O)=C3C(C4=C(O)C=CC(N(C)C)=C4C[C@@]3([H])C[C@@]2([H])[C@@H]1N(C)C)=O)=O.Cl
493.94
Research
COVID-19-immunoregulation