missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Minocycline Hydrochloride, MP Biomedicals™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Color | Yellow |
Quantity | 250 mg |
Formula Weight | 493.9 |
Physical Form | Powder |
Chemical Name or Material | Minocycline Hydrochloride |
Safety and Handling
Product Identifier
- Minocycline Hydrochloride
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Minocycline Hydrochloride
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction