missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Minocycline hydrochloride, Tocris Bioscience™
Click to view available options
Quantity:
50 mg
Description
Minocycline hydrochloride is a tetracycline antibiotic. Displays neuroprotective, antiapoptotic, anti-inflammatory and antimicrobial effects. Acts as a matrix metalloproteinase (MMP) inhibitor; attenuates disease severity in mouse models of multiple sclerosis. Orally active and brain penetrant. Allows formation of extended pluripotent stem (EPS) cells in combination with CHIR 99021 (Cat.No. 4423), (S)-(+)-Dimethindene maleate (Cat.No. 1425) and human leukemia inhibitory factor.
For more information about how Minocycline hydrochloride may be used, see our protocol: Generation and Propagation of EPS cells (LCDM Cocktail) .
Specifications
Specifications
CAS | 13614-98-7 |
Molecular Formula | C23H28ClN3O7 |
MDL Number | MFCD00083669 |
Quantity | 50 mg |
Synonym | minocycline hydrochloride, minocycline hcl, minomycin, minocin, minocycline chloride, arestin, vectrin, minomax, periocline, tri-mino |
InChI Key | KDLQIOPKJDNQIM-WUURTAMISA-N |
SMILES | [H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@@H]3CC4=C(C=CC(O)=C4C(O)=C3C(=O)[C@]2(O)C(=O)\C(=C(\N)O)C1=O)N(C)C |
IUPAC Name | hydrogen (2Z,4S,4aS,5aR,12aS)-2-[amino(hydroxy)methylidene]-4,7-bis(dimethylamino)-10,11,12a-trihydroxy-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride |
Molecular Weight (g/mol) | 493.94 |
PubChem CID | 54685925 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction