missing translation for 'onlineSavingsMsg'
Learn More
Mirtazapine, 98%, Thermo Scientific Chemicals Product Code.: 15355058

Mirtazapine, 98%, Thermo Scientific Chemicals

Product Code. 15355058
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15355058

Brand: Thermo Scientific Acros 456170050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85650-52-8
Molecular Formula C17H19N3
Molecular Weight (g/mol) 265.35
InChI Key RONZAEMNMFQXRA-UHFFFAOYSA-N
Synonym mirtazapine, remeron, mepirzepine, remergil, zispin, remergon, rexer, 6-azamianserin, remeron soltab, mepirzapin
PubChem CID 4205
ChEBI CHEBI:6950
SMILES CN1CCN2C(C1)C3=CC=CC=C3CC4=C2N=CC=C4

Specifications

Melting Point 121.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 5 g
Formula Weight 265.35
Percent Purity 98%
Chemical Name or Material Mirtazapine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467636 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number456170050,456170010
Lot NumberA0464736 Certificate TypeCertificate of Analysis Date24/07/2024 Catalog Number456170050,456170010
Lot NumberA0406229 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog Number456170050,456170010
Lot NumberA0350375 Certificate TypeCertificate of Analysis Date04/03/2022 Catalog Number456170050,456170010
Product Identifier
  • Mirtazapine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mirtazapine, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.