missing translation for 'onlineSavingsMsg'
Learn More
Mirtazapine, Tocris Bioscience™ Product Code.: 30001439

Mirtazapine, Tocris Bioscience™

Product Code. 30001439
10 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 30001439

Brand: Tocris Bioscience 2018/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent 5-HT2 antagonist; also 5-HT3, H1 and α2 antagonist; antidepressant

Mirtazapine is an antidepressant agent; potent 5-HT2, 5-HT3 and histamine H1 receptor antagonist and moderately potent α2-adrenoceptor antagonist (pKi values are 8.05, ∼ 8.1, 9.3 and 6.95 respectively). Enhances noradrenaline (NA) release in rat brain via inhibition of α2-adrenergic autoreceptors and displays only weak affinity for monoamine transporters (pKi values are 5.6, < 5 and < 5.1 for inhibition of NA, dopamine and 5-HT uptake respectively). Increases hippocampal NA and 5-HT levels in rats following systemic administration in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85650-52-8
Molecular Formula C17H19N3
Molecular Weight (g/mol) 265.36
InChI Key RONZAEMNMFQXRA-UHFFFAOYSA-N
Synonym mirtazapine, remeron, mepirzepine, remergil, zispin, remergon, rexer, 6-azamianserin, remeron soltab, mepirzapin
PubChem CID 4205
ChEBI CHEBI:6950
SMILES CN1CCN2C(C1)C3=CC=CC=C3CC4=C2N=CC=C4

Specifications

Quantity 10 mg
Formula Weight 265.36
Percent Purity >99%
Chemical Name or Material Mirtazapine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mirtazapine, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.