Learn More
Schmidlin™ Mistral Evolution Nitrogen Generator
This generator produces a continuous supply of high purity nitrogen.
Brand:  Schmidlin™ DB-EVO-25-EU
Encompass_Preferred
 View more versions of this product
Product Code. 17728879
  • 23726.82 € / Each
Explore more special offers
Description
Description
  • Increases efficiency and return on investment
  • On-demand supply 24/7
  • Flow rate at 8 barg (116 psig)
  • LC/MS grade purity
  • Integrated, low noise, oil-free compressors
  • Proprietary carbon molecular sieve technology
  • Complete 'plug and play' LC/MS solution
Specifications
Specifications
83.5 cm
68 cm
8 bar
48.5 cm
110/120 V at 60 Hz, 220/240 V at 50 Hz
1800 W
25 L/min.
35°C
19 in.
Outlet: 6 mm OD Compression or 1/4 in.
>99%