missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Mithramycin A, 90%, from Streptomyces argillaceus

Product Code. 10599213
Click to view available options
Quantity:
1 mg
5 mg
25 mg
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18378-89-7
Molecular Formula C52H76O24
Molecular Weight (g/mol) 1085.156
MDL Number MFCD00135618
InChI Key CFCUWKMKBJTWLW-ZUZMCERRSA-N
PubChem CID 122172942
IUPAC Name (2R,3R)-2-[(2R,4S,5S,6S)-4-[(2R,4S,5S,6S)-4-[(2R,4R,5R,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-5-hydroxy-6-methyloxan-2-yl]oxy-3-[(1R,3R,4S)-3,4-dihydroxy-1-methoxy-2-oxopentyl]-6-[(2S,4S,5S,6S)-4-[(2S,4S,5R,6S)-4,5-dih
SMILES CC1C(C(CC(O1)OC2CC(OC(C2O)C)OC3=CC4=CC5=C(C(=O)C(C(C5)C(C(=O)C(C(C)O)O)OC)OC6CC(C(C(O6)C)O)OC7CC(C(C(O7)C)O)OC8CC(C(C(O8)C)O)(C)O)C(=C4C(=C3C)O)O)O)O

Specifications

Type Mithramycin A
Physical Form Powder
CAS Min % 88.0
CAS Max % 100.0
Melting Point 180.0°C to 183.0°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Merck Index 15, 7652
Packaging Glass Bottle
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in lower alcohols, acetone, ethyl acetate, moderatly soluble in chloroform, slightly soluble in ether, benzene
Formula Weight 1085.16
Percent Purity 90%
Color Brown to Red
Quantity 1 mg
Chemical Name or Material Mithramycin A, 90%
Show More Show Less
compliance-icons
Product Identifier
  • Mithramycin A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H311-Toxic in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'