missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MitoSOX™ Mitochondrial Superoxide Indicators, for live-cell imaging

MitoSOX superoxide indicators are novel fluorogenic dyes specifically targeted to mitochondria in live cells. Oxidation of the MitoSOX reagent by mitochondrial superoxide produces bright green or red fluorescence.

144.00 € - 618.00 €

Specifications

Detection Method Fluorescence
Product Line MitoSOX™
View More Specs

Products 5
Product Code Brand Quantity Excitation/Emission Indicator Color Price Quantity & Availability  
Product Code Brand Quantity Excitation/Emission Indicator Color Price Quantity & Availability  
17837181
View Documents
Invitrogen™
M36005
1 vial x 9 μg ∼488/510 nm MitoSox Green Green
144.00 €
9µg
Please to purchase this item. Need a web account? Register with us today!
17845110
View Documents
Invitrogen™
M36006
5 vials x 9 μg ∼488/510 nm MitoSox Green Green
374.00 €
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
11579096
View Documents
Invitrogen™
M36008
10 vials x 50 μg ∼396/610 nm MitoSox Red Red
618.00 €
500µg
Please to purchase this item. Need a web account? Register with us today!
17847181
View Documents
Invitrogen™
M36007
1 vial x 50 μg ∼396/610 nm MitoSox Red Red
144.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
17866721
View Documents
Invitrogen™
M36009
2 Vials (1 Green, 1 Red) 488/510 nm, 396/610 nm MitoSox Green, MitoSox Red Green, Red
235.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description
Show More
Specifications

Specifications

Fluorescence
MitoSOX™
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ MitoSOX™ Mitochondrial Superoxide Indicators, for live-cell imaging

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.