Learn More
Mixed Calibration Standard 2A For Environmental Analysis, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
298.00 €
Specifications
Chemical Name or Material | Mixed Calibration Standard 2A |
---|---|
Packaging | Triple-leached LDPE bottle |
Concentration or Composition Notes | Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. |
Technique | AA, ICP |
For Use With (Equipment) | AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
K | 200μg/mL |
Mo | 100μg/mL |
Na | 100μg/mL |
Li | 50μg/mL |
Sr | 10μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Mixed Calibration Standard 2A | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
5% HNO3 | |
Components in 5% HNO3: Li •50μg/mL Mo •100μg/mL K •200μg/mL Na •100μg/mL Sr •10μg/mL |
Triple-leached LDPE bottle | |
AA, ICP | |
Certified Reference Material | |
Environmental |
Safety and Handling
Causes severe skin burns and eye damage
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.