missing translation for 'onlineSavingsMsg'
Learn More

Mixed Calibration Standard 4 For Environmental Analysis, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

192.00 € - 319.00 €

Specifications

Chemical Name or Material Mixed Calibration Standard 4
Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Grade Environmental
View More Specs

Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
11405928
View Documents
SPEX Certiprep
MIXSTD4-100
125 mL Triple-leached LDPE bottle
192.00 €
125mL
Please to purchase this item. Need a web account? Register with us today!
11415928
View Documents
SPEX Certiprep
MIXSTD4-500
500 mL Triple-leached LDPE bottle
319.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ca1000μg/mL
K400μg/mL
Al200μg/mL
Na200μg/mL
Cr20μg/mL
Ni20μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Mixed Calibration Standard 4
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Environmental
5% HNO3
Triple-leached LDPE bottle
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Components in 5% HNO3:
Al    •200μg/mL
Ca    •1000μg/mL
Cr    •20μg/mL
Ni    •20μg/mL
K    •400μg/mL
Na    •200μg/mL
Videos
Safety and Handling

Safety and Handling

Danger: Causes severe skin burns and eye damage Do not breathe dusts or mists. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. If in eyes, rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.

missing translation for 'dotInformation' : Hazardous

Recommended Storage : Ambient

Product Identifier
  • Mixed Calibration Standard 4

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P363-Wash contaminated clothing before reuse.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : nitric acid

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only