missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IKA™ Lab Dancer Vortex Mixer

Use this attractively designed, test tube shaker for mixing small test samples. The IKA™ Lab Dancer Vortex Mixer is ideal for small containers up to 30mm in diameter such as test tubes, centrifuge tubes, Eppendorf beakers.
Brand: IKA™ 0003365000
267.74 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Small, compact and reliable- For small containers up to 30mm in diameter, e.g. test tubes, centrifuge tubes, Eppendorf™ beakers
- Fixed speed (2,800rpm)
- Excellent mixing action
- The upper casing and the test tube surface are made from inert plastic. The bottom section is a coated, zinc die cast casing
- Includes light 12V power pack set
- Orbital shaking motion
- 100% ON time permissable

Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
IKA™ Lab Dancer Vortex Mixer > for test tubes and centrifuge tubes up to 30mm dia 230V 50Hz 100mm dia x 70m height 2,800rpm fixed speed IKA
Spot an opportunity for improvement?Share a Content Correction