Learn More
MK-2894, MedChemExpress
MedChemExpress MK-2894 is a potent, selective, orally active and high affinity (Ki=0.56 nM) full antagonist against E prostanoid receptor 4 (EP4 receptor) (IC50=2.5 nM). MK-2894 possesses potent anti-inflammatory activity in animal models of pain/inflammation and can be used for the research of arthritis.

222.75 € - 1645.65 €
Specifications
Chemical Name or Material | MK-2894 |
---|---|
Physical Form | Solid |
CAS | 1006036-87-8 |
Color | Off-White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16473958
![]() |
Medchem Express
HY-10413 |
10 mg |
390.15 €
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16456677
![]() |
Medchem Express
HY-10413 |
5 mg |
222.75 €
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16430317
![]() |
Medchem Express
HY-10413 |
50 mg |
1645.65 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MK-2894 is a potent, selective, orally active and high affinity (Ki=0.56 nM) full antagonist against E prostanoid receptor 4 (EP4 receptor) (IC50=2.5 nM). MK-2894 possesses potent anti-inflammatory activity in animal models of pain/inflammation and can be used for the research of arthritis.Specifications
MK-2894 | |
1006036-87-8 | |
Research | |
COVID-19-immunoregulation | |
O=C(O)C1=CC=C(C2(NC(C3=C(C)SC(C)=C3CC4=CC=C(C(F)(F)F)C=C4)=O)CC2)C=C1 | |
473.51 | |
Research |
Solid | |
Off-White | |
C25H22F3NO3S | |
DMSO : ≥ 50 mg/mL (105.59 mM) | |
473.51 | |
95.14% |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.