missing translation for 'onlineSavingsMsg'
Learn More

(-)-MK 801 maleate, Tocris Bioscience™

Product Code. 30001372
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

NMDA antagonist; less active enantiomer of (+)-MK 801

Less active enantiomer (10-fold less potent in behavioral studies) than (+)-MK 801 maleate.

(+)-enantiomer also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121917-57-5
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.375
InChI Key QLTXKCWMEZIHBJ-FWHYOZOBSA-N
Synonym --mk 801 maleate, unii-ef13o02iv0, dizocilpine maleate, --mk-801 hydrogen maleate, dizocilpine maleate,-, -mk-801 hydrogen maleate, --mk 801 hydrogen maleate, --mk-801 hydrogen maleate hplc, 5r,10s---5-methyl-10,11-dihydro-5h-dibenzo a,d cylcohepten-5,10-imine maleate
PubChem CID 16219612
SMILES CC12C3=CC=CC=C3CC(N1)C4=CC=CC=C24.C(=CC(=O)O)C(=O)O

Specifications

Quantity 10 mg
Formula Weight 337.37
Percent Purity >99%
Chemical Name or Material (-)-MK 801 maleate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.